Learn More
Chemglass Life Sciences Filter Flask, 2L, 40/35, Plastic Coated
SDP
Supplier:  Chemglass Life Sciences CG142404
Encompass_Preferred
2L filter flask for use with the CG142401 filtration support assembly. Plastic coated. Recommended for filtering corrosive solutions, particulate removal from HPLC solvents, and filtration analysis with solid phase extraction disks.
Catalog No. CG142404