Learn More
Ace Glass, Inc. Flask, reaction/storage, 50mL, outer 14/20 standard taper joint0-3 inlet valve side arm, No-Air, Schlenk
SDP
Supplier:  Ace Glass, Inc. 7754209
Encompass_Preferred
Flask, reaction/storage, 50mL, outer 14/20 standard taper joint0-3 inlet valve side arm, No-Air, Schlenk
Catalog No. 50-975-372