Learn More
Ace Glass, Inc. Flask, round bottom, 50mL, (2)14/20 necks, 2mm glass stopcock sidearm, No-Air
SDP
Supplier:  Ace Glass, Inc. 779902
Encompass_Preferred
50mL No-Air round bottom flask with (2) 14/20 standard taper necks, and a 2mm glass stopcock sidearm. Excellent flask for reactions, and No-Air or vacuum applications.
Catalog No. 50-979-464