Learn More
Ace Glass, Inc. 4 liter dewar flask, cs/2, sp/1
SDP
Supplier:  Ace Glass, Inc. 1254009
Encompass
4L dewar flask molded of high-density polyethylene, with expanded polystyrene cover, and polyethylene coated bail-type handle. Suitable for cryogenic work. Double walls will withstand temps from -196 to 100 C. Vented, insulating cover.
Catalog No. NC0346327