Learn More
Wilmad Labglass FLASK ERL 125ML ST 22 STPR
Supplier:  Wilmad Labglass LG7790106
Encompass_Preferred
Erlenmeyer Flask, w/ Standard Taper Outer Flask Joint. Capacity: 125mL. Flask Joint Size: 22. Supplied complete w/ Glass pennyhead stopper
Catalog No. 50-803-494