Learn More
Hach Company Flask, Erlenmeyer, Polymethylpentene Capacity 250 mL, 4/pk
Supplier:  Hach Company 2089876
Encompass
Polymethylpentene flask with polypropylene screwcap features clarity, chemical resistance, strength, and autoclavability (Do not engage threads during cycle). Useful for titrations, preparation and storage of media.
Catalog No. 50-199-5316