Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemglass Life Sciences Chemglass Erlenmeyer flask, 2,000 mL, GL-45 thread
SDP

Supplier:  Chemglass Life Sciences CG154423

Encompass_Preferred

The Chemglass 2,000 mL Erlenmeyer flask features a GL-45 screw thread top and is supplied complete with a polypropylene linerless closure and pouring ring. It is graduated to approximate volumes and packaged individually for laboratory use.

  • Capacity: 2,000 mL.
  • Thread type: GL-45 screw thread.
  • Closure: Polypropylene linerless closure with pouring ring.
  • Graduated to approximate volumes.
  • Packaging: Supplied individually.
  • Overall height: Approximately 275 mm.

Catalog No. 50-273-3582


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.