Learn More
Chemglass Life Sciences Chemglass Erlenmeyer Flask, 250 mL, Polycarbonate, Baffled Bottom, Seal Cap, Sterile
SDP
Supplier:  Chemglass Life Sciences CGN2093250
Encompass_Preferred
Sterile 250 mL polycarbonate Erlenmeyer flask with a quadruple-baffled bottom and a seal cap designed for mixing, shaking, and anaerobic applications. Non-pyrogenic, DNase/RNase-free, and sterilized by E-beam (SAL=10); graduations are molded for accuracy and each unit is individually packaged in a sterile bag.
  • Polycarbonate construction.
  • 250 mL capacity with molded graduations.
  • Quadruple baffled bottom for improved mixing.
  • Seal cap included for anaerobic applications.
  • Sterile - E-beam sterilized (SAL=10).
  • Non-pyrogenic and DNase/RNase-free.
  • Individually packaged in sterile bags.
Catalog No. 50-273-3478