Learn More
Ace Glass, Inc. Flask, Erlenmeyer, 25mL, 14/10 with 20-400 external thread, supplied with cap
SDP
Supplier:  Ace Glass, Inc. 947144
Encompass_Preferred
25mL microscale Erlenmeyer flask with a 14/10, 20-400 externally threaded joint. Supplied with a holed cap, a PTFE-faced silicone septum, and a FETFE o-ring.
Catalog No. 50-976-901