Learn More
Chemglass Life Sciences Chemglass Erlenmeyer Flask, 2 L - High-Efficiency Polycarbonate, Baffled, Sterile
SDP
Supplier:  Chemglass Life Sciences CGN21662L
Encompass_Preferred
A 2 L high-efficiency Erlenmeyer flask with a baffled bottom, designed for cell culture and bioprocess applications. Constructed from medical-grade polycarbonate for high transparency and durability, it is supplied sterile and supports gas exchange when used with vented caps.
  • High-efficiency baffled bottom for improved aeration during shaking.
  • Medical-grade polycarbonate construction with clear graduation scale for easy volume observation.
  • Supplied sterile (E-beam sterilized) and non-pyrogenic; typically packaged four per case.
  • Vent cap option with 0.22 μm breathable film to allow gas exchange while preventing contamination.
  • Compatible with USP Class VI and ISO 10993 / USP <661> material standards; suitable for applications requiring biocompatible plastics.
Catalog No. 50-273-3447