Learn More
Chemglass Life Sciences Chemglass Erlenmeyer Flask, 2 L High-Efficiency Polycarbonate, Baffled Bottom, Sterile (Nest)
SDP
Supplier:  Chemglass Life Sciences CGN21662LSC
Encompass_Preferred
A 2 L high-efficiency Erlenmeyer flask made from medical-grade polycarbonate for sterile cell culture, shaking, and general laboratory mixing applications. The baffled bottom improves aeration and mixing while clear material and printed graduations allow easy visual monitoring. Supplied sterile in nest packaging for aseptic handling.
  • Polycarbonate construction meets USP Class VI.
  • Baffled bottom promotes consistent solution agitation.
  • Clear graduation scale for easy observation.
  • Sterilized by e-beam and provided non-pyrogenic.
  • Withstands temperatures up to 121 °C.
  • Packaged sterile in nest form, 4 per case.
Catalog No. 50-273-3446