Learn More
Chemglass Life Sciences Erlenmeyer flask, 3 L, wide mouth, high-efficiency polycarbonate, baffled bottom, sterile, seal cap
SDP
Supplier:  Chemglass Life Sciences CGN21663WSC
Encompass_Preferred
High-efficiency 3 L Erlenmeyer flask made from transparent polycarbonate for laboratory mixing and culture applications. The wide-mouth, baffled-bottom design improves aeration and mixing; the flask is supplied sterile and fitted with a seal cap for aseptic handling.
  • 3 L capacity.
  • Polycarbonate construction for high transparency.
  • Wide mouth for easy filling and cleaning.
  • Baffled bottom to improve mixing and aeration.
  • Supplied sterile; USP Class VI compliance.
  • Seal cap included.
Catalog No. 50-273-3442