Learn More
Chemglass Life Sciences Chemglass 3 L Wide-Mouth High-Efficiency Polycarbonate Erlenmeyer Flask, Baffled Bottom, Sterile, Vent Filter Cap (CGN-2166-3W)
SDP
Supplier:  Chemglass Life Sciences CGN21663W
Encompass_Preferred
High-efficiency 3 L wide-mouth polycarbonate Erlenmeyer flask designed for laboratory mixing and cell culture applications. The baffled bottom improves aeration and mixing, and the vent filter cap allows gas exchange while maintaining sterility.
  • High-efficiency wide-mouth design for improved oxygen transfer and mixing.
  • Polycarbonate construction provides clear visibility and impact resistance.
  • Baffled bottom promotes better aeration and agitation during shaking.
  • Sterile and e-beam sterilized for immediate use.
  • Vent filter cap with 0.22 μm breathable filter maintains sterility while permitting gas exchange.
  • Packaged in sterile nests for clean handling; 4 per case.
Catalog No. 50-273-3443