Learn More
Chemglass Life Sciences Chemglass High Efficiency Erlenmeyer Flask, 3 L, Polycarbonate, Baffled, Seal Cap (CGN-2166-3LSC)
SDP
Supplier:  Chemglass Life Sciences CGN21663LSC
Encompass_Preferred
High-efficiency 3 L Erlenmeyer flask made from medical-grade polycarbonate, designed for shaken culture and laboratory mixing. Baffled bottom improves agitation and mixing performance, and the clear graduation scale enables easy volume observation. Sterilized by E-beam and packaged in sterile bags; four flasks per case.
  • Medical-grade polycarbonate (USP Class VI) for high transparency and impact resistance.
  • Baffled bottom for improved and consistent agitation on flask shakers.
  • Seal cap provides a secure, non-vented closure.
  • E-beam sterilized and non-pyrogenic.
  • Autoclavable up to 121 °C.
  • Packaged sterile, 4 per case.
  • Recommended working volume 30-40% and starting shaker speed 75-125 RPM.
Catalog No. 50-273-3444