Learn More
Chemglass Life Sciences Chemglass Erlenmeyer Flask, 3 L, High Efficiency Polycarbonate, Baffled Bottom, Sterile (CGN-2166-3L)
SDP
Supplier:  Chemglass Life Sciences CGN21663L
Encompass_Preferred
High-efficiency 3 L Erlenmeyer flask made from medical-grade polycarbonate for culture and mixing applications. The baffled bottom improves mixing and aeration, and the clear external graduation scale enables easy volume observation.
  • Medical-grade polycarbonate construction.
  • 3 L nominal capacity with clear external graduation scale.
  • Baffled bottom for enhanced mixing and aeration.
  • Sterile and USP Class VI compliant.
  • Sterilized by E-Beam and non-pyrogenic.
  • Packaged in sterile nests, 4 per case.
  • Withstands temperatures up to 121°C.
Catalog No. 50-273-3445