Learn More
Chemglass Life Sciences Chemglass Erlenmeyer Flask, 500 mL Polycarbonate, Sterile, Seal Cap (Nest, Pack of 12)
SDP
Supplier:  Chemglass Life Sciences CGN2090500
Encompass_Preferred
Sterile 500 mL polycarbonate Erlenmeyer flask supplied nest-packed and individually bagged. The seal cap provides secure closure for mixing, shaking, and storage; molded graduations aid volume measurement. Sterilized by e-beam and certified DNase/RNase-free.
  • 500 mL capacity.
  • Polycarbonate construction for impact resistance.
  • Seal cap for secure mixing and storage.
  • Sterile; E-beam sterilized; SAL=10.
  • Nest-packed and individually bagged; pack quantity: 12 (C12).
  • Molded graduations for measurement accuracy.
  • Non-pyrogenic and DNase/RNase-free.
  • Dimensions: height 175.8 mm; neck diameter 39 mm; bottom diameter 101 mm.
Catalog No. 50-273-3483