Learn More
Chemglass Life Sciences Flask, Erlenmeyer, 5L, High Efficiency, Polycarbonate, Plain Bottom, with Pre-Assembled Tube System, Sterile
SDP
Supplier:  Chemglass Life Sciences CGN21675L
Encompass_Preferred
High-efficiency, large-volume culture flasks allow cells to show strong viability with large expression amount of proteins in the culture of mammalian cells and insect cells. During the culturing process, the use rate of the shaker is significantly increased, and the survival rate and viability of cells are both dramatically elevated.culture flasks also provide high repeatability, which allows highly inter-batch consistency of cell growth and yield. Features: PC bottle, which meets the USP Class 6 Standard, with high transparency, has strong impact resistance, oxidation resistance and can withstand a high temperature up to 121 degrees C. There’s a scale made by injection molding on the flask body to facilitate observing the filled liquid volume. The wide-mouth neck design is convenient for sampling and liquid filling. DNase/RNase free, no pyrogens. Sterilized by E-Beam. Packaged in sterile bags
Catalog No. 50-273-3440