Learn More
Chemglass Life Sciences Flask, Fernbach, 2L, Polycarbonate, Plain Bottom, 70mm Vented Cap, Max Volume: 2, 400mL
SDP
Supplier:  Chemglass Life Sciences CLS2170FP2L
Encompass_Preferred
Erlenmeyer and Fernbach culture flasks are designed for shaker and cell culture applications. All flasks feature a 70mm neck size that provides easy access for filling and dispensing. The unique shape of the Fernbach flask provides an increased surface area to volume ratio for improved gas exchange. The flasks are available in 2L Erlenmeyer, 2L and 3L Fernbach sizes, with plain bottoms. The polypropylene caps are knurled for secure grip and are configured in 70mm large vent or non-vented styles. The flasks are constructed from an optically clear polycarbonate material that can be autoclaved. All flasks include a vented cap and are individually packed in bags and gamma irradiation sterilized. Additional caps are available separately.
Catalog No. 50-000-83289