Learn More
Ace Glass, Inc. Flask, half-jacketed, three neck, 3L, 29/42 center, (2) 24/40sides, #11 Ace-Thred connections on inlet/outlets, for use with 3/8in ID tubing
SDP
Supplier:  Ace Glass, Inc. 6945355
Encompass
Flask, half-jacketed, three neck, 3L, 29/42 center, (2) 24/40sides, #11 Ace-Thred connections on inlet/outlets, for use with 3/8in ID tubing
Catalog No. 50-978-981