Learn More
Bellco Glass, Inc. Flask Locking Tray,20x20
SDP
Supplier:  Bellco Glass, Inc. 774320020
Encompass_Preferred
Includes 5 dividers Size: 20x20". MADE IN THE USA. How many size flasks will fit on the tray:125ml Erlenmeyer flasks – 36ea250ml Erlenmeyer flasks – 25ea300ml Erlenmeyer flasks – 25ea500ml Erlenmeyer flasks – 16ea1000mL Erlenmeyer flasks – 9ea2000mL Erlenmeyer flasks – 9ea
Catalog No. 50-194-6127