Learn More
Chemglass Life Sciences Chemglass recovery flask, 1,000 mL heavy-wall wide-mouth single neck (GL80)
SDP
Supplier:  Chemglass Life Sciences CG1512X11
Encompass_Preferred
This 1,000 mL heavy-wall recovery flask is designed for vacuum and rotary evaporation applications. The wide-mouth, single-neck design with GL80 thread allows easy emptying and access, while the balanced contour supports controlled evaporation of foaming solvents.
  • Heavy wall construction suitable for vacuum applications.
  • Wide-mouth single neck with GL80 thread for easy filling and cleaning.
  • Flask contour promotes controlled evaporation of foaming solvents during rotary evaporation.
  • Large opening permits easy access with a spatula or policeman.
  • Made from glass for chemical resistance and thermal stability.
Catalog No. 50-273-3591