Learn More
Ace Glass, Inc. Flask, pear-shaped, 5mL, 14/10 cap seal joint
SDP
Supplier:  Ace Glass, Inc. 947760
Encompass_Preferred
5mL pear-shaped, single neck, heavy duty flask with a 14/10 cap-seal joint. Cap-seal flasks are used in the ACE microscale chemistry product line, and feature a 14/10 ground joint with external threads. Supplied with a holed cap, a PTFE-faced septum, and a FETFE o-ring.
Catalog No. 50-976-922