Learn More
Ace Glass, Inc. Flask, pear-shaped, 5mL, 14/20 ground joint
SDP
Supplier:  Ace Glass, Inc. 947702
Encompass_Preferred
5mL pear-shaped, single neck, heavy duty flask with a 14/20 ground joint.
Catalog No. 50-976-908