Learn More
Wilmad Labglass FLASK RB 1N 1000ML 45/50
Supplier:  Wilmad Labglass LG7220242
Encompass_Preferred
Boiling Flask, Round Bottom, Short Neck, 1000mL capacity, with [ST] 45/50 outer joint.
Catalog No. 50-807-563