Learn More
Wilmad Labglass FLASK RB 1N 100ML 19/22
Supplier:  Wilmad Labglass ML1110772
Encompass_Preferred
Boiling Flask, Round Bottom, Short Neck, 100mL capacity, with [ST] 19/22 outer joint.
Catalog No. 50-807-736