Learn More
Wilmad Labglass FLASK RB 1N 12000ML 55/50
Supplier:  Wilmad Labglass LG7220270
Encompass_Preferred
Boiling Flask, Round Bottom, Short Neck, 12L capacity, with [ST] 55/50 outer joint.
Catalog No. 50-806-449