Learn More
Wilmad Labglass FLASK RB 1N 2000ML 24/40
Supplier:  Wilmad Labglass LG7220156
Encompass_Preferred
Boiling Flask, Round Bottom, Short Neck, 2000mL capacity, with [ST] 24/40 outer joint.
Catalog No. 50-807-220