Learn More
Wilmad Labglass FLASK RB 1N 3000ML 35/25
Supplier:  Wilmad Labglass LG7240138
Encompass
Boiling Flask, Round Bottom, Short Neck, 3000mL capacity, with [SJ] 35/25 socket joint.
Catalog No. 50-804-362