Learn More
Wilmad Labglass FLASK RB 3000ML 24/40C V24/40S
Supplier:  Wilmad Labglass LG7290270
Encompass_Preferred
Boiling Flask, Round Bottom, 2-Neck, 3000mL capacity. Joint Sizes: Center Neck [ST] 24/40 outer; Vertical Side Neck: [ST] 24/40 outer. Round bottom two neck boiling flask with outer joints on both the center neck and the vertical side neck.
Catalog No. 50-803-614