Learn More
Wilmad Labglass FLASK RECOVERY 100ML 14/20
Supplier:  Wilmad Labglass ML1155706
Encompass_Preferred
Boiling Flask, Recovery, 100mL Capacity with [ST] 14/20 Outer Joint. Upper portion of the flask has a gradual taper making it easier to remove reaction products. Lower hemisphere fits comparably sized heating mantles.
Catalog No. 50-802-914