Learn More
Wilmad Labglass FLASK RECOVERY 2000ML 24/40
Supplier:  Wilmad Labglass LG7245110
Encompass_Preferred
Recovery Flask, 2000mL capacity, with [ST] 24/40 outer joint. Upper portion of the flask has a gradual taper making it easier to remove reaction products. Lower hemisphere fits comparably sized heating mantles.
Catalog No. 50-807-037