Learn More
Wilmad Labglass FLASK RECOVERY 25ML 24/40
Supplier:  Wilmad Labglass LG7245098
Encompass_Preferred
Recovery Flask, 25mL capacity, with [ST] 24/40 outer joint. Upper portion of the flask has a gradual taper making it easier to remove reaction products. Lower hemisphere fits comparably sized heating mantles.
Catalog No. 50-803-359