Learn More
Ace Glass, Inc. Flask, recovery, 500mL, 29/32, indented, Buchi; 00452
SDP
Supplier:  Ace Glass, Inc. 399410
Encompass_Preferred
500mL replacement recovery flask for Buchi 00452 rotary evaporator. Flask is indented, pear shaped with a heavy wall, and features a 29/32 standard taper single neck.
Catalog No. 50-971-534