Learn More
Wilmad Labglass FLASK RECOVERY 50ML 24/40
Supplier:  Wilmad Labglass LG7245100
Encompass_Preferred
Recovery Flask, 50mL capacity, with [ST] 24/40 outer joint. Upper portion of the flask has a gradual taper making it easier to remove reaction products. Lower hemisphere fits comparably sized heating mantles.
Catalog No. 50-803-031
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.