Learn More
Chemglass Life Sciences Flask, Round Bottom, 25mL, Heavy Wall, 3-Neck, 19/22 CN - 14/20 SN's Angled 20°
SDP
Supplier:  Chemglass Life Sciences CG1524A03
Encompass_Preferred
Flask, Round Bottom, 25mL, Heavy Wall, 19/22 - 14/20, 3-Neck, Angled 20¿.Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 19/22Side neck: 14/20
Catalog No. 50-873-132