Learn More
Chemglass Life Sciences Chemglass Round-bottom Flask, 5-Neck Heavy Wall, 5,000 mL (CG-1535-08)
SDP
Supplier:  Chemglass Life Sciences CG153508
Encompass_Preferred
The Chemglass CG-1535-08 is a heavy-wall, five-neck round-bottom flask with a 5,000 mL capacity, designed for multi-connection reaction assemblies and laboratory distillation setups. It features a larger center joint and vertical side necks with standard taper joints to accept common glass fittings and adapters.
  • 5,000 mL capacity for medium- to large-scale reactions.
  • Heavy-wall construction for improved durability and thermal stress resistance.
  • Five necks to allow simultaneous attachments and probe access.
  • Center neck uses a 45/50 standard taper joint; side necks use 24/40 standard taper joints.
  • Vertical neck orientation for compact, vertical assemblies.
  • Suitable for multi-neck reaction, distillation, and reflux applications.
Catalog No. 50-273-3583