Learn More
Chemglass Life Sciences Chemglass Flask, Round Bottom, 500 mL, 3-Neck Heavy-Wall with Drain Valve, 24/40 Joints (CG-1523-V-01)
SDP
Supplier:  Chemglass Life Sciences CG1523V01
Encompass_Preferred
Heavy-wall, three-neck, 500 mL round-bottom flask with a lower outlet drain valve for controlled draining and complete agitation of contents. Standard taper outer joints on the center and side necks provide secure connections for laboratory assemblies.
  • Capacity: 500 mL.
  • Three necks with standard taper (24/40) joints.
  • Heavy-wall construction for improved durability.
  • Lower outlet drain valve enables controlled draining of contents.
  • Drain valve sidearm accepts 1/2 in Swagelok fittings.
Catalog No. 50-273-3586