Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, single neck, 100mL, 35/25
SDP
Supplier:  Ace Glass, Inc. 6902235
Encompass_Preferred
100mL heavy wall spherical receiving flask. Flask has a short neck and a 35/25 outer joint.
Catalog No. 50-973-563