Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, two neck, 3L, 29/42 center, 29/42 side
SDP
Supplier:  Ace Glass, Inc. 6927258
Encompass_Preferred
3L two neck, round bottom heavy wall flask with standard joints. Center neck and angled side neck both have 29/42 joints.
Catalog No. 50-973-588