Learn More
Chemglass Life Sciences Flask, Shake, 500mL, 4 Baffles, Constriction, Delong Neck, Approx OD x Height (mm): 100 x 200
SDP
Supplier:  Chemglass Life Sciences CLS205004
Encompass_Preferred
Flask, Shake, 500mL, 4 Baffles, Constriction, Delong Neck, Approx OD x Height (mm): 100 x 200. Flask has 4 bottom baffles and a constriction just below the neck to prevent splashing from reaching the closure.
Catalog No. 50-153-2273