Learn More
Chemglass Life Sciences Flask Support Ring, Rubber, Black
SDP
Supplier:  Chemglass Life Sciences CG305601
Encompass_Preferred
Flask Support Ring, Rubber, Black. Flask support ring has a concave recess on top to support round bottom flasks from 250 to 500mL. 5-1/8” diameter.
Catalog No. 50-131-1209