Learn More
Chemglass Life Sciences 200mL Volumetric Flask, Class A, #16 Outer Stopper Neck, PE Stopper
SDP
Supplier:  Chemglass Life Sciences CG160206
Encompass_Preferred
200mL Volumetric Flask, Class A, #16 Outer Stopper Neck, PE Stopper. Flasks are manufactured to conform to Federal Specifications NNN-F-289 for Type 1, Class A unserialized flasks and ASTM E-288. Supplied complete with one closed bottom polyethylene stopper. For additional stoppers, see CG-8740. NOTE: THE CG-1602-20 CONFORMS TO ISO 1042, DIN 12664 CLASS A FLASKS AND IS A DURAN PRODUCT.
Catalog No. 50-153-1187