Learn More
Chemglass Life Sciences 5mL Volumetric Flask, Class A, #9 Outer Stopper Neck, Glass Stopper
SDP
Supplier:  Chemglass Life Sciences CG160001
Encompass_Preferred
5mL Volumetric Flask, Class inAin, #9 Outer Stopper Neck, Glass Stopper. Flasks are manufactured to conform to Federal Specifications NNN-F-289 for Type 1, Class ¿A¿ unserialized flasks and ASTM E-288. Supplied complete with one solid glass pennyhead stopper. For additional stoppers, see CG-3018.
Catalog No. CG160001