Learn More
Bellco Glass, Inc. Flat Bottom AHB Spinner Complete, 1000mL
SDP
Supplier:  Bellco Glass, Inc. 196701000
Encompass_Preferred
•Designed for the efficient growth of suspension cells at low speeds. MADE IN THE USA•Flask Specifications:o1000 mL flat bottom, Type 1 Class A BorosilicateoDimensions: 135mm OD x 240mm OAHoTwo 45mm Sidearms angled 60º offset centeroOne 100mm Screw thread center neckoGraduations: 100mL to 1000mL, every 100mLoIncludes two 45mm blue PP screw caps and pouring rings•Impeller Assembly Specifications:oConsists of PTFE coated magnet and white Teflon magnet holder•Glass Shaft Specifications:oType 1 Class A BorosilicateoDimensions: 7mm OD x 230mm long•Cap Assembly Specifications:oConsists of one 100mm blue PP screw cap with 16.2 mm hole for black compression fitting, 100mm white silicone liner with 15.8mm hole, 7mm black compression fitting
Catalog No. 50-194-5140