Learn More
Medchemexpress LLC Flavanone | 487-26-3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN712810MM/1ML
Encompass_Preferred
Flavanone is a naturally occurring flavone with inhibitory activity for human estrogen synthetase (aromatase) and acts as an inhibitor for the ERK/p38/NF-κB signaling pathway. It exhibits oral activity and antitumor efficacy, demonstrating its potential in various biological processes.
  • Inhibits cell proliferation, migration, and invasion of A549 cells
  • Arrests cell cycle at G1 phase in vitro
  • Inhibits expression of MMP-2, u-PA, cyclin D1, and CDK2
  • Promotes generation of TIMP-2 and PAI-1
  • Exhibits antitumor efficacy in C57BL/6J mouse models
  • Limits tumor growth and inhibits metastasis of LLC cells in vivo
Catalog No. 50-004-18175