Learn More
Medchemexpress LLC Flavin adenine dinucleotide | 146-14-5 | 98.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1654R100MG
Encompass_Preferred
Flavin adenine dinucleotide (FAD) is a redox cofactor, specifically a prosthetic group of a protein, involved in several important enzymatic reactions in metabolism. This product is the analytical standard of Flavin adenine dinucleotide and is intended for research and analytical applications.
  • Analytical standard of flavin adenine dinucleotide.
  • Intended for research and analytical applications.
  • Functions as a redox cofactor.
  • Involved in important enzymatic reactions in metabolism.
Catalog No. 50-004-13199