Learn More
AdipoGen -Flavipucine
SDP
Supplier:  AdipoGen BVT0279M001
Encompass_Preferred
Chemical. CAS 72597-14-9. Formula C12H15NO4. MW 237.3. Isolated from Phoma sp. BS 7204. Phytotoxin Fruit rot toxin. Antibiotic. Antibacterial and antifungal compound.
Catalog No. 50-256-2022