Learn More
ATCC Streptomyces avermitilis
Supplier:  ATCC 31267
Encompass
Streptomyces avermitilis strain NRRL 8165 [JCM 5070, MA-4680, NCIMB 12804] is a bacterial type strain that was isolated from soil in Itoh City, Japan. This whole-genome sequenced culture produces C-076 and derivatives, including avermectins.
Catalog No. 50-238-1658