Learn More
Cayman Chemical FlavokawaIn 1I 10mg
SDP
Supplier:  Cayman Chemical 3939410 MG
Encompass_Preferred
A flavokawain derivative with anticancer and antiviral activities; inhibits the proliferation of gefitinib-resistant H1975 NSCLC cells by 36%, as well as decreases protein levels of EGFR, c-Met, HER2, Akt, and Cdk4 and increases protein levels of Hsp70, markers of Hsp90 inhibition, at 30 µM; intramuscular administration reduces viral titers in PRRSV-infected pigs
Catalog No. 50-291-8646