Learn More
Selleck Chemical LLC Flavokawain A
SDP
Supplier:  Selleck Chemical LLC S560025MG
Encompass_Preferred
Flavokawain A extracted from kava is an apoptotic inducers and anticarcinogenic agent Flavokawain A can down-regulation of antiapoptotic proteins such as XIAP survivin and Bcl-xL thereby changing the balance between apoptotic and antiapoptotic molecules and then induce cell death in tumor cells
Catalog No. 50-000-03461